Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminoethanol, 99%, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
20 L
Packaging:
Amber Glass Bottle
Glass Bottle
Poly Drum
141-43-5
141-43-5
Packaging Amber Glass Bottle
Refractive Index 1.453-1.455
Quantity 4 L
CAS Max % 1
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.