Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminoethyl methacrylate hydrochloride, 90%, stabilized, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2420-94-2
C6H11NO2·HCl
165.62
MFCD00078260
XSHISXQEKIKSGC-UHFFFAOYSA-N
2-aminoethyl methacrylate hydrochloride, 2-aminoethylmethacrylate hydrochloride, 2-aminoethyl 2-methylacrylate hydrochloride, methacrylic acid, 2-aminoethyl ester, hydrochloride, polymers, 2-aminoethyl 2-methylprop-2-enoate hydrochloride, 2-propenoic acid, 2-methyl-, 2-aminoethyl ester, hydrochloride 1:1, 2-propenoic acid, 2-methyl-, 2-aminoethyl ester, hydrochloride, aminoethylmethacrylate, acmc-209wgm, timtec-bb sbb003905
75495
2-aminoethyl 2-methylprop-2-enoate;hydrochloride
CC(=C)C(=O)OCCN.Cl
2420-94-2
C6H11NO2·HCl
165.62
MFCD00078260
XSHISXQEKIKSGC-UHFFFAOYSA-N
2-aminoethyl methacrylate hydrochloride, 2-aminoethylmethacrylate hydrochloride, 2-aminoethyl 2-methylacrylate hydrochloride, methacrylic acid, 2-aminoethyl ester, hydrochloride, polymers, 2-aminoethyl 2-methylprop-2-enoate hydrochloride, 2-propenoic acid, 2-methyl-, 2-aminoethyl ester, hydrochloride 1:1, 2-propenoic acid, 2-methyl-, 2-aminoethyl ester, hydrochloride, aminoethylmethacrylate, acmc-209wgm, timtec-bb sbb003905
75495
2-aminoethyl 2-methylprop-2-enoate;hydrochloride
CC(=C)C(=O)OCCN.Cl
Color Brown to White
Packaging Glass bottle
Melting Point 116.0°C to 118.0°C
Infrared Spectrum Authentic
Quantity 25g
Stabilizer 500 to 1000ppm phenothiazine
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 88% min. (Argentometry)
Solubility Information Solubility in water: soluble
Formula Weight 165.62
Physical Form Crystalline Powder
Percent Purity 90%
Chemical Name or Material 2-Aminoethyl methacrylate hydrochloride, 90%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 219-343-4

Recommended Storage : Refrigerator +4°C