Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminoindan-2-carboxylic acid hydrate, 97%, ACROS Organics™

Click to view available options
Quantity:
250mg
Packaging:
Glass bottle
97%
C10H11NO2·xH2O
MFCD03419294
2-Aminoindan-2-carboxylic acid hydrate, 97%
97%
C10H11NO2·xH2O
MFCD03419294
2-Aminoindan-2-carboxylic acid hydrate, 97%
Color Beige
Packaging Glass bottle
Quantity 250mg
Water 11% max. (K.F.)
Infrared Spectrum Authentic
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethanol
Molecular Weight (g/mol) 177.2
Formula Weight 177.2
Physical Form Powder
Percent Purity 97%

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

Recommended Storage : Refrigerator +4°C