Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminoindan-2-carboxylic Acid Hydrochloride, 97%, ACROS Organics™

Click to view available options
Quantity:
250mg
Packaging:
Glass bottle
33584-60-0
C10H11NO2·ClH
213.66
MFCD00800587
TUBGNZQKMUMUEA-UHFFFAOYSA-N
2-amino-2,3-dihydro-1h-indene-2-carboxylic acid hydrochloride,2-aminoindan-2-carboxylic acid hydrochloride,2-amino-1,3-dihydroindene-2-carboxylic acid hydrochloride,1h-indene-2-carboxylicacid, 2-amino-2,3-dihydro-, hydrochloride 1:1
45789447
2-amino-1,3-dihydroindene-2-carboxylic acid;hydrochloride
C1C2=CC=CC=C2CC1(C(=O)O)N.Cl
33584-60-0
C10H11NO2·ClH
213.66
MFCD00800587
TUBGNZQKMUMUEA-UHFFFAOYSA-N
2-amino-2,3-dihydro-1h-indene-2-carboxylic acid hydrochloride,2-aminoindan-2-carboxylic acid hydrochloride,2-amino-1,3-dihydroindene-2-carboxylic acid hydrochloride,1h-indene-2-carboxylicacid, 2-amino-2,3-dihydro-, hydrochloride 1:1
45789447
2-amino-1,3-dihydroindene-2-carboxylic acid;hydrochloride
C1C2=CC=CC=C2CC1(C(=O)O)N.Cl
Melting Point 285°C to 289°C
Packaging Glass bottle
Quantity 250mg
Infrared Spectrum Authentic
Color White to Beige
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 213.66
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-Aminoindan-2-carboxylic acid hydrochloride
Show More Show Less