Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Aminoindan 99.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
| Color | Brown |
| Boiling Point | 229°C |
| Quantity | 25 g |
| UN Number | 2735 |
| Formula Weight | 133.19 |
| Percent Purity | ≥99.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Aminoindan |
Safety and Handling
| TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction