Alfa Aesar™ 2-Aminoindane, 97%

Manufacturer: Alfa Aesar™ H31023-06

Quantity 5g

 View more versions of this product

Catalog No. AAH3102306

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 2975-41-9
    Molecular Formula C9H11N
    Formula Weight 133.19
    MDL Number MFCD00082597
    UN Number UN2922
    Chemical Name or Material 2-Aminoindane

    Specifications

    Formula Weight 133.19
    Molecular Formula C9H11N
    MDL Number MFCD00082597
    UN Number UN2922
    Quantity 5g
    Flash Point 100°C (212°F)
    Odor Amine-like
    Health Hazard 2 GHS H Statement
    H301-H314-H318
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Boiling Point 229°C
    CAS 2975-41-9
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
    Chemical Name or Material 2-Aminoindane
    Assay 97%

    GHS H Statement
    H301-H314-H318
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit