Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Aminomethyl)pyridine, 99%

Click to view available options
Quantity:
5g
25g
100g
3731-51-9
C6H8N2
108.144
MFCD00006360
WOXFMYVTSLAQMO-UHFFFAOYSA-N
2-picolylamine,2-aminomethyl pyridine,2-pyridinemethanamine,2-aminomethylpyridine,2-pyridinemethylamine,2-pyridylmethylamine,2-picolinamine,2-pyridinemethaneamine,2-pyridylmethyl amine,pyridine, 2-aminomethyl
19509
CHEBI:81387
pyridin-2-ylmethanamine
C1=CC=NC(=C1)CN
3731-51-9
C6H8N2
108.144
MFCD00006360
WOXFMYVTSLAQMO-UHFFFAOYSA-N
2-picolylamine,2-aminomethyl pyridine,2-pyridinemethanamine,2-aminomethylpyridine,2-pyridinemethylamine,2-pyridylmethylamine,2-picolinamine,2-pyridinemethaneamine,2-pyridylmethyl amine,pyridine, 2-aminomethyl
19509
CHEBI:81387
pyridin-2-ylmethanamine
C1=CC=NC(=C1)CN
Applications
2-(Amino methyl)pyridine is a synthesis reagent used in the synthesis of PPD analogues. It is also used in the preparation of 5-nitro-2-furancarboxylamides displaying potent tryoanocidal activity against trypanosomiasis.

Solubility
Soluble in water (1000 mg/ml at 20°C), and ethanol.

Notes
Air sensitive. Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents, acids, air.
Boiling Point 202°C to 204°C
Melting Point -20°C
Quantity 5g
Odor Amine-like
pH 11 to 12
Flash Point 90°C (194°F)
UN Number UN2735
Beilstein 108054
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material 2-(Aminomethyl)pyridine
Show More Show Less

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

RTECSNumber : US1840000

TSCA : TSCA