Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Aminomethylindane, 97%

Click to view available options
Quantity:
1g
5g
146737-65-7
C10H13N
147.221
MFCD04114560
COWVOJUAZGKGRQ-UHFFFAOYSA-N
2-aminomethyl-indane,indan-2-yl-methylamine,2,3-dihydro-1h-inden-2-yl methanamine,indan-2-yl-methylamine hydrochloride,2-aminomethylindane,2-aminomethyl indan,indan-2-ylmethanamine,indan-2-ylmethylamine,2-aminomethyl indane,1h-indene-2-methanamine, 2,3-dihydro
22346315
2,3-dihydro-1H-inden-2-ylmethanamine
C1C(CC2=CC=CC=C21)CN
146737-65-7
C10H13N
147.221
MFCD04114560
COWVOJUAZGKGRQ-UHFFFAOYSA-N
2-aminomethyl-indane,indan-2-yl-methylamine,2,3-dihydro-1h-inden-2-yl methanamine,indan-2-yl-methylamine hydrochloride,2-aminomethylindane,2-aminomethyl indan,indan-2-ylmethanamine,indan-2-ylmethylamine,2-aminomethyl indane,1h-indene-2-methanamine, 2,3-dihydro
22346315
2,3-dihydro-1H-inden-2-ylmethanamine
C1C(CC2=CC=CC=C21)CN
Quantity 1g
Beilstein 3241648
Formula Weight 147.22
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Aminomethylindane

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger