Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Aminonicotinamide, 97%

Click to view available options
Quantity:
1g
5g
13438-65-8
C6H7N3O
137.142
MFCD00128873
HTPCDVLWYUXWQR-UHFFFAOYSA-N
2-aminonicotinamide,2-amino-nicotinamide,2-amino nicotinamide,3-pyridinecarboxamide,2-amino,3-pyridinecarboxamide, ar-amino,2-amino-3-pyridinecarboxamide,amino-nicotinamide,ar-aminonicotinamide,ar-amino-3-pyridinecarboxamide,nicotinamide, amino
506623
2-aminopyridine-3-carboxamide
C1=CC(=C(N=C1)N)C(=O)N
13438-65-8
C6H7N3O
137.142
MFCD00128873
HTPCDVLWYUXWQR-UHFFFAOYSA-N
2-aminonicotinamide,2-amino-nicotinamide,2-amino nicotinamide,3-pyridinecarboxamide,2-amino,3-pyridinecarboxamide, ar-amino,2-amino-3-pyridinecarboxamide,amino-nicotinamide,ar-aminonicotinamide,ar-amino-3-pyridinecarboxamide,nicotinamide, amino
506623
2-aminopyridine-3-carboxamide
C1=CC(=C(N=C1)N)C(=O)N
Melting Point 194°C to 200°C
Quantity 1g
Color Yellow
UN Number UN2811
Beilstein 3613
Formula Weight 137.14
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Aminonicotinamide

GHS H Statement
H302-H319-H317
Harmful if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P305+P351+P338-P301+P312-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.