Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminophenol Hydrochloride 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
51-19-4
C6H8ClNO
145.59
MFCD00035471
QUDZAFKCASPJLX-UHFFFAOYSA-N
5800
hydrogen 2-aminophenol chloride
[H+].[Cl-].NC1=CC=CC=C1O
51-19-4
C6H8ClNO
145.59
MFCD00035471
QUDZAFKCASPJLX-UHFFFAOYSA-N
5800
hydrogen 2-aminophenol chloride
[H+].[Cl-].NC1=CC=CC=C1O
Color Undesignated
Quantity 25g
Formula Weight 145.59
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,N)
Chemical Name or Material 2-Aminophenol Hydrochloride

RTECSNumber : SJ6069000

TSCA : No

Recommended Storage : Refrigerator