Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminophenyl Phenyl Sulfide 98.0+%, TCI America™

Click to view available options
Quantity:
25g
100g
500g
1134-94-7
C12H11NS
201.287
DGBISJKLNVVJGD-UHFFFAOYSA-N
2-Aminodiphenyl Sulfide, 2-(Phenylthio)aniline
365393
2-phenylsulfanylaniline
C1=CC=C(C=C1)SC2=CC=CC=C2N
1134-94-7
C12H11NS
201.287
DGBISJKLNVVJGD-UHFFFAOYSA-N
2-Aminodiphenyl Sulfide, 2-(Phenylthio)aniline
365393
2-phenylsulfanylaniline
C1=CC=C(C=C1)SC2=CC=CC=C2N
Color Undesignated
Melting Point 43°C
Quantity 100g
Boiling Point 258°C
UN Number 3077
Formula Weight 201.29
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Aminophenyl Phenyl Sulfide

EINECSNumber : (3)-4188

TSCA : No