Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Aminophenylacetonitrile, 97%

Click to view available options
Quantity:
1g
5g
2973-50-4
C8H8N2
132.166
MFCD00091082
LMDPYYUISNUGGT-UHFFFAOYSA-N
2-2-aminophenyl acetonitrile,2-aminophenylacetonitrile,2-aminobenzyl cyanide,2-aminophenyl acetonitrile,2-aminobenzylcyanide,o-aminobenzyl cyanide,o-aminophenylacetonitrile,2-aminobenzeneacetonitrile,2-2-aminophenyl ethanenitrile,benzeneacetonitrile, 2-amino
76307
2-(2-aminophenyl)acetonitrile
C1=CC=C(C(=C1)CC#N)N
2973-50-4
C8H8N2
132.166
MFCD00091082
LMDPYYUISNUGGT-UHFFFAOYSA-N
2-2-aminophenyl acetonitrile,2-aminophenylacetonitrile,2-aminobenzyl cyanide,2-aminophenyl acetonitrile,2-aminobenzylcyanide,o-aminobenzyl cyanide,o-aminophenylacetonitrile,2-aminobenzeneacetonitrile,2-2-aminophenyl ethanenitrile,benzeneacetonitrile, 2-amino
76307
2-(2-aminophenyl)acetonitrile
C1=CC=C(C(=C1)CC#N)N
Melting Point 69°C to 72°C
Quantity 5g
Color White
UN Number UN3439
Beilstein 2716483
Formula Weight 132.17
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 2-Aminophenylacetonitrile

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.