Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminophenylarsonic Acid 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
2045-00-3
C6H8AsNO3
217.056
MFCD00007630
LQCOCUQCZYAYQK-UHFFFAOYSA-N
o-arsanilic acid,2-aminobenzenearsonic acid,o-aminobenzenearsonic acid,2-aminophenyl arsonic acid,arsenical,unii-4p7t2ph7dl,2-arsanilic acid,o-aminophenylarsonic acid,4p7t2ph7dl,arsonic acid,as-2-aminophenyl
73161
(2-aminophenyl)arsonic acid
C1=CC=C(C(=C1)N)[As](=O)(O)O
2045-00-3
C6H8AsNO3
217.056
MFCD00007630
LQCOCUQCZYAYQK-UHFFFAOYSA-N
o-arsanilic acid,2-aminobenzenearsonic acid,o-aminobenzenearsonic acid,2-aminophenyl arsonic acid,arsenical,unii-4p7t2ph7dl,2-arsanilic acid,o-aminophenylarsonic acid,4p7t2ph7dl,arsonic acid,as-2-aminophenyl
73161
(2-aminophenyl)arsonic acid
C1=CC=C(C(=C1)N)[As](=O)(O)O
Color Undesignated
Melting Point 153°C
Quantity 25g
UN Number 3465
Formula Weight 217.06
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material 2-Aminophenylarsonic Acid