Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminophenylboronic acid hydrochloride, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
863753-30-4
C6H8BNO2·ClH
173.41
WPDASZCYRKGSTO-UHFFFAOYSA-N
2-aminophenylboronic acid hydrochloride,2-aminophenyl boronic acid hydrochloride,2-aminophenylboronic acid hcl,2-aminophenylboronic acid, hydrochloride,2-2-aminophenyl boronic acid hydrochloride,boronic acid, 2-aminophenyl-, hydrochloride,pubchem1746,ksc914i9f,2-aminophenylboronic acid,hcl,2-aminophenylboronic acid, hcl
16427090
(2-aminophenyl)boronic acid;hydrochloride
B(C1=CC=CC=C1N)(O)O.Cl
863753-30-4
C6H8BNO2·ClH
173.41
WPDASZCYRKGSTO-UHFFFAOYSA-N
2-aminophenylboronic acid hydrochloride,2-aminophenyl boronic acid hydrochloride,2-aminophenylboronic acid hcl,2-aminophenylboronic acid, hydrochloride,2-2-aminophenyl boronic acid hydrochloride,boronic acid, 2-aminophenyl-, hydrochloride,pubchem1746,ksc914i9f,2-aminophenylboronic acid,hcl,2-aminophenylboronic acid, hcl
16427090
(2-aminophenyl)boronic acid;hydrochloride
B(C1=CC=CC=C1N)(O)O.Cl
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 96% min. (HPLC)
Formula Weight 173.41
Percent Purity 97%
Chemical Name or Material 2-Aminophenylboronic acid hydrochloride

GHS Signal Word: Warning