Learn More
2-Aminopyridine, 99%, Thermo Scientific Chemicals
Description
A compound has been shown to reversibly block voltage-dependent potassium channels. Also a common impurity from the synthesis of compounds found in hair dye. Used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics. It is used as a research tool, in characterizing subtypes of the potassium channel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
A compound has been shown to reversibly block voltage-dependent potassium channels. Also a common impurity from the synthesis of compounds found in hair dye. Used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics. It is used as a research tool, in characterizing subtypes of the potassium channel.
Solubility
Slightly soluble in water(8g/100 mL at 19°C)
Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.
Specifications
Melting Point | 56°C to 60°C |
Boiling Point | 209°C to 211°C |
Flash Point | 92°C (197°F) |
Quantity | 1000 g |
UN Number | UN2671 |
Beilstein | 105785 |
Sensitivity | Hygroscopic |
Merck Index | 14,473 |
Solubility Information | Slightly soluble in water(8g/100 mL at 19°C) |
Formula Weight | 94.12 |
Show More |
Safety and Handling
Hazard Category | H301-H312-H315-H319-H500 |
Hazard Statement | GHS H Statement H301-H331-H312-H315-H319-H335 Toxic if swallowed. Toxic if inhaled. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: AMINOPYRIDINES |
EINECSNumber | 207-988-4 |
RTECSNumber | US1575000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.