Alfa Aesar™ 2-Aminopyridine, 99%

Manufacturer: Alfa Aesar™ A1237414

Quantity 25g

 View more versions of this product

Catalog No. AAA1237414

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 504-29-0
    Molecular Formula C5H6N2
    Formula Weight 94.12
    MDL Number MFCD00006312
    UN Number UN2671
    Beilstein 105785
    Merck Index 14,473
    Synonym 2-Pyridinamine; 2-Pyridylamine
    Chemical Name or Material 2-Aminopyridine

    Specifications

    Formula Weight 94.12
    Health Hazard 3 GHS P Statement
    P280h-P273-P270-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    Avoid release to the environment.
    Do not eat, drink or smoke when using this product.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Boiling Point 209° to 211°C
    Molecular Formula C5H6N2
    MDL Number MFCD00006312
    CAS 504-29-0
    Merck Index 14,473
    Synonym 2-Pyridinamine; 2-Pyridylamine
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: AMINOPYRIDINES
    Melting Point 56° to 60°C
    UN Number UN2671
    Beilstein 105785
    Health Hazard 1 Danger
    RTECS Number US1575000
    TSCA TSCA
    Quantity 25g
    Flash Point 92°C (197°F)
    Health Hazard 2 GHS H Statement
    H301-H331-H312-H315-H319-H335
    Toxic if swallowed.
    Toxic if inhaled.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 2-Aminopyridine
    Assay 99%

    Description

    Applications
    A compound has been shown to reversibly block voltage-dependent potassium channels. Also a common impurity from the synthesis of compounds found in hair dye. Used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics. It is used as a research tool, in characterizing subtypes of the potassium channel.

    Solubility
    Slightly soluble in water(8g/100 mL at 19°C)

    Notes
    Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.

    GHS H Statement
    H301-H331-H312-H315-H319-H335
    Toxic if swallowed.
    Toxic if inhaled.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P273-P270-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    Avoid release to the environment.
    Do not eat, drink or smoke when using this product.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit