Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Aminopyrimidine-5-boronic acid, 95%

Click to view available options
Quantity:
250mg
1g
5g
936250-22-5
C4H6BN3O2
138.921
MFCD07375147
CGHYQZASLKERLV-UHFFFAOYSA-N
2-aminopyrimidine-5-boronic acid,2-aminopyrimidin-5-yl boronic acid,2-amino-pyrimidine-5-boronic acid,2-amino-5-pyrimidineboronic acid,2-aminopyrimidin-5-yl-5-boronic acid,2-amino-5-boronopyrimidine,acmc-209rmr,2-aminopyrimidine 5-boronic acid,2-amino-5-pyrimidinylboronic acid,2-amino-pyrimidin-5-ylboronic acid
17750211
(2-aminopyrimidin-5-yl)boronic acid
B(C1=CN=C(N=C1)N)(O)O
936250-22-5
C4H6BN3O2
138.921
MFCD07375147
CGHYQZASLKERLV-UHFFFAOYSA-N
2-aminopyrimidine-5-boronic acid,2-aminopyrimidin-5-yl boronic acid,2-amino-pyrimidine-5-boronic acid,2-amino-5-pyrimidineboronic acid,2-aminopyrimidin-5-yl-5-boronic acid,2-amino-5-boronopyrimidine,acmc-209rmr,2-aminopyrimidine 5-boronic acid,2-amino-5-pyrimidinylboronic acid,2-amino-pyrimidin-5-ylboronic acid
17750211
(2-aminopyrimidin-5-yl)boronic acid
B(C1=CN=C(N=C1)N)(O)O
Applications
2-Aminopyrimidine-5-boronic acid is used as pharmaceutical intermediates.

Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents.
Melting Point 206°C to 211°C
Quantity 250mg
Beilstein 15758675
Formula Weight 138.92
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-Aminopyrimidine-5-boronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C