Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Aminothiazole-4-carboxamide, 95%

Click to view available options
Quantity:
250mg
1g
5g
118452-02-1
C4H5N3OS
143.164
MFCD08689593
YSNYFPFEBBRSHS-UHFFFAOYSA-N
2-aminothiazole-4-carboxamide,2-amino-thiazole-4-carboxylamide,4-thiazolecarboxamide, 2-amino,2-amino-thiazole-4-carboxylic acid amide,2-aminothiazole-4-carboxylamide,4-thiazolecarboxamide,2-amino,acmc-20agwj,2-aminothiazol-4-carboxamide,ksc510m7r,2-amino-thiazole-4-carboxamide
257149
2-amino-1,3-thiazole-4-carboxamide
C1=C(N=C(S1)N)C(=O)N
118452-02-1
C4H5N3OS
143.164
MFCD08689593
YSNYFPFEBBRSHS-UHFFFAOYSA-N
2-aminothiazole-4-carboxamide,2-amino-thiazole-4-carboxylamide,4-thiazolecarboxamide, 2-amino,2-amino-thiazole-4-carboxylic acid amide,2-aminothiazole-4-carboxylamide,4-thiazolecarboxamide,2-amino,acmc-20agwj,2-aminothiazol-4-carboxamide,ksc510m7r,2-amino-thiazole-4-carboxamide
257149
2-amino-1,3-thiazole-4-carboxamide
C1=C(N=C(S1)N)C(=O)N
Quantity 5g
Beilstein 4969494
Formula Weight 143.16
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-Aminothiazole-4-carboxamide