Alfa Aesar™ 2-Aminothiazole, 97%

Manufacturer: Alfa Aesar™ A1202630

Quantity 250g

 View more versions of this product

Catalog No. AAA1202630

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 96-50-4
    Molecular Formula C3H4N2S
    Formula Weight 100.14
    MDL Number MFCD00005325
    Beilstein 105738
    Merck Index 14,479
    Chemical Name or Material 2-Aminothiazole

    Specifications

    Formula Weight 100.14
    Health Hazard 3 GHS P Statement
    P281-P264-P301+P312-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Wash thoroughly after handling.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00005325
    CAS 96-50-4
    Boiling Point 117°C (15mmHg)
    Beilstein 105738
    Merck Index 14,479
    Molecular Formula C3H4N2S
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H341-H302
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Melting Point 88° to 92°C
    Quantity 250g
    RTECS Number XJ2100000
    TSCA TSCA
    Chemical Name or Material 2-Aminothiazole
    Assay 97%

    Description

    Solubility
    Soluble in water, alcohol and ether.

    Notes
    Incompatible with strong oxidizing agents, strong acids, acid chlorides, acid anhydrides, nitric acid and sulfuric acid.

    GHS H Statement
    H341-H302
    Suspected of causing genetic defects.
    Harmful if swallowed.

    GHS P Statement
    P281-P264-P301+P312-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Wash thoroughly after handling.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit