Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Aminothiazole hydrochloride, 97%

Click to view available options
Quantity:
100g
500g
6149-03-7
C14H21NaO3S
292.37
MFCD00270826
ASJUTVUXOMPOQH-UHFFFAOYSA-M
sodium 4-n-octylbenzenesulfonate,sodium 4-octylbenzenesulfonate,unii-hts5em67p9,hts5em67p9,4-octylbenzenesulfonic acid sodium salt,4-n-octylbenzenesulfonic acid sodium salt,acmc-2097it,dsstox_cid_27594,dsstox_rid_82441,dsstox_gsid_47594
23670838
sodium;4-octylbenzenesulfonate
CCCCCCCCC1=CC=C(C=C1)S(=O)(=O)[O-].[Na+]
6149-03-7
C14H21NaO3S
292.37
MFCD00270826
ASJUTVUXOMPOQH-UHFFFAOYSA-M
sodium 4-n-octylbenzenesulfonate,sodium 4-octylbenzenesulfonate,unii-hts5em67p9,hts5em67p9,4-octylbenzenesulfonic acid sodium salt,4-n-octylbenzenesulfonic acid sodium salt,acmc-2097it,dsstox_cid_27594,dsstox_rid_82441,dsstox_gsid_47594
23670838
sodium;4-octylbenzenesulfonate
CCCCCCCCC1=CC=C(C=C1)S(=O)(=O)[O-].[Na+]
Quantity 100g
Beilstein 5404572
Formula Weight 136.6
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Aminothiazole hydrochloride