Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Aminothiophenol, 98%

Click to view available options
Quantity:
25g
100g
500g
137-07-5
C6H7NS
125.189
MFCD00007702
VRVRGVPWCUEOGV-UHFFFAOYSA-N
2-aminothiophenol,o-aminothiophenol,2-mercaptoaniline,o-mercaptoaniline,o-aminobenzenethiol,2-mercaptaniline,benzenethiol, 2-amino,2-amino thiophenol,1-amino-2-mercaptobenzene,2-aminobenzene-1-thiol
8713
2-aminobenzenethiol
C1=CC=C(C(=C1)N)S
137-07-5
C6H7NS
125.189
MFCD00007702
VRVRGVPWCUEOGV-UHFFFAOYSA-N
2-aminothiophenol,o-aminothiophenol,2-mercaptoaniline,o-mercaptoaniline,o-aminobenzenethiol,2-mercaptaniline,benzenethiol, 2-amino,2-amino thiophenol,1-amino-2-mercaptobenzene,2-aminobenzene-1-thiol
8713
2-aminobenzenethiol
C1=CC=C(C(=C1)N)S
Applications
2-Aminothiophenol was used in the synthesis of 1,5-benzothiazepines derivatives by reacting with 1,3-diphenylpropenone derivatives and using aluminosilicate solid catalysts. It was also used in the synthesis of benzothiazole2 and 3-(benzothiazol-2-yl)coumarin derivatives. The ternary complexes of copper (II) with salicylidene-2-aminothiophenol (L) and glycine, alanine, valine and histidene amino acids.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
Refractive Index 1.642
Density 1.17
Sensitivity Air Sensitive
Flash Point 79°C (174°F)
Quantity 500g
Boiling Point 234°C
Odor Stench
Melting Point 18°C to 23°C
UN Number UN3267
Beilstein 606076
Solubility Information Insoluble in water.
Formula Weight 125.19
Percent Purity 98%
Chemical Name or Material 2-Aminothiophenol
Show More Show Less

GHS H Statement
H301-H314-H318-H373-H227-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
May cause an allergic skin reaction.

P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H302-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 205-277-3

RTECSNumber : DC0600000

TSCA : Yes

Recommended Storage : Ambient temperatures