Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Azidomethyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific™

Click to view available options
Quantity:
1g
5g
1223598-41-1
C13H18BN3O2
259.116
MFCD22124732
DWSJHXRRSHEEOE-UHFFFAOYSA-N
2-2-azidomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, amtb515, 2-azidomethyl benzeneboronic acid pinacol ester
57416890
2-[2-(azidomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2CN=[N+]=[N-]
1223598-41-1
C13H18BN3O2
259.116
MFCD22124732
DWSJHXRRSHEEOE-UHFFFAOYSA-N
2-2-azidomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, amtb515, 2-azidomethyl benzeneboronic acid pinacol ester
57416890
2-[2-(azidomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2CN=[N+]=[N-]
Quantity 5g
Formula Weight 260.12
Percent Purity 95%
Chemical Name or Material 2-(Azidomethyl)benzeneboronic acid pinacol ester

TSCA : No

Recommended Storage : Ambient temperatures