Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzothienylboronic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
98437-23-1
C8H7BO2S
178.02
MFCD01075674
YNCYPMUJDDXIRH-UHFFFAOYSA-N
2-benzothienylboronic acid,benzo b thiophen-2-ylboronic acid,benzo b thiophene-2-boronic acid,benzothiophene-2-boronic acid,1-benzothien-2-ylboronic acid,thianaphthene-2-boronic acid,benzo b thiophene-2-ylboronic acid,2-benzothiopheneboronic acid,benzothiophen-2-ylboronic acid
2359
1-benzothiophen-2-ylboronic acid
B(C1=CC2=CC=CC=C2S1)(O)O
98437-23-1
C8H7BO2S
178.02
MFCD01075674
YNCYPMUJDDXIRH-UHFFFAOYSA-N
2-benzothienylboronic acid,benzo b thiophen-2-ylboronic acid,benzo b thiophene-2-boronic acid,benzothiophene-2-boronic acid,1-benzothien-2-ylboronic acid,thianaphthene-2-boronic acid,benzo b thiophene-2-ylboronic acid,2-benzothiopheneboronic acid,benzothiophen-2-ylboronic acid
2359
1-benzothiophen-2-ylboronic acid
B(C1=CC2=CC=CC=C2S1)(O)O
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 25g
Color White
Melting Point 236.0°C to 237.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 178.02
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 2-Benzothienylboronic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.