Alfa Aesar™ 2-Benzoylpyridine, 99%

Manufacturer: Alfa Aesar™ A1085718

Quantity 50g

 View more versions of this product

Catalog No. AAA1085718

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 91-02-1
    Molecular Formula C12H9NO
    Formula Weight 183.21
    MDL Number MFCD00006300
    Beilstein 120283
    Synonym Phenyl 2-pyridyl ketone
    Chemical Name or Material 2-Benzoylpyridine

    Specifications

    Formula Weight 183.21
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C12H9NO
    CAS 91-02-1
    TSCA TSCA
    MDL Number MFCD00006300
    Beilstein 120283
    Synonym Phenyl 2-pyridyl ketone
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Boiling Point 313°C
    Quantity 50g
    Health Hazard 1 Warning
    Melting Point 41° to 45°C
    Flash Point 167°C (332°F)
    RTECS Number OB6400000
    Chemical Name or Material 2-Benzoylpyridine
    Assay 99%

    Description

    Applications
    The Schiff bases(2-Benzoylpyridine ) and copper(II) complexes display moderate antifungal activities. 2-Benzoylpyridine is an aromatic substituent which has selective antiproliferative effects, with the majority having greater antineoplastic activity. Reaction of 2-benzoylpyridine N(4)-phenyl thiosemicarbazone (HL) with a variety of copper salts yielded a series of copper(II) complexes of the type [CuLX]. Complexes of 2-benzoylpyridine thiosemicarbazone (H2Bz4DH) as well as of its N(4)-methyl (H2Bz4Me) and N(4)-phenyl (H2Bz4Ph) analogues with iron(II), nickel(II) and zinc(II) were characterized by a variety of spectroscopic techniques.

    Solubility
    Soluble in methanol. Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit