Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzyl-1,3-dioxolane, 94%, Acros Organics

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
101-49-5
C10H12O2
164.2
MFCD00003215
SSZACLYPEFCREM-UHFFFAOYSA-N
2-benzyldioxolan,1,3-dioxolane, 2-phenylmethyl,phenylacetaldehyde glycol acetal,phenylacetaldehyde ethylene acetal,1,3-dioxolane, 2-benzyl,phenylacetaldehyde ethyleneglycol acetal,phenylacetaldehyde ethylene glycol acetal,1,3-dioxolane,2-phenylmethyl,benzyldioxolan,1, 2-benzyl
7562
2-benzyl-1,3-dioxolane
C1COC(O1)CC2=CC=CC=C2
101-49-5
C10H12O2
164.2
MFCD00003215
SSZACLYPEFCREM-UHFFFAOYSA-N
2-benzyldioxolan,1,3-dioxolane, 2-phenylmethyl,phenylacetaldehyde glycol acetal,phenylacetaldehyde ethylene acetal,1,3-dioxolane, 2-benzyl,phenylacetaldehyde ethyleneglycol acetal,phenylacetaldehyde ethylene glycol acetal,1,3-dioxolane,2-phenylmethyl,benzyldioxolan,1, 2-benzyl
7562
2-benzyl-1,3-dioxolane
C1COC(O1)CC2=CC=CC=C2
Specific Gravity 1.085
Boiling Point 115°C to 120°C (12.0mmHg)
Flash Point 108°C
Density 1.0850g/mL
Refractive Index 1.5200 to 1.522
Color Colorless to Yellow
Infrared Spectrum Authentic
Quantity 100g
Packaging Glass bottle
CAS Min % 92.0
CAS Max % 100.0
Assay Percent Range 92% min. (GC)
Solubility Information Solubility in water: insoluble
Formula Weight 164.2
Physical Form Liquid
Percent Purity 94%
Chemical Name or Material 2-Benzyl-1, 3-dioxolane
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 202-946-1