Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Benzyl-2-imidazoline hydrochloride, 99%

Click to view available options
Quantity:
25g
100g
59-97-2
C10H13ClN2
196.678
MFCD00012693
RHTNTTODYGNRSP-UHFFFAOYSA-N
tolazoline hydrochloride,tolazoline hcl,2-benzyl-4,5-dihydro-1h-imidazole hydrochloride,arterodyl,tolavad,tolpal,priscoline hydrochloride,tolazoline chloride,imidaline hydrochloride,benzazoline hydrochloride
6048
2-benzyl-4,5-dihydro-1H-imidazole;hydrochloride
C1CN=C(N1)CC2=CC=CC=C2.Cl
59-97-2
C10H13ClN2
196.678
MFCD00012693
RHTNTTODYGNRSP-UHFFFAOYSA-N
tolazoline hydrochloride,tolazoline hcl,2-benzyl-4,5-dihydro-1h-imidazole hydrochloride,arterodyl,tolavad,tolpal,priscoline hydrochloride,tolazoline chloride,imidaline hydrochloride,benzazoline hydrochloride
6048
2-benzyl-4,5-dihydro-1H-imidazole;hydrochloride
C1CN=C(N1)CC2=CC=CC=C2.Cl
Quantity 100g
Melting Point 174°C to 176°C
Merck Index 14,9506
Formula Weight 196.68
Percent Purity 99%
Chemical Name or Material 2-Benzyl-2-imidazoline hydrochloride

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA