Alfa Aesar™ 2-Benzyl-4,4-dimethyl-2-oxazoline, 95%

Manufacturer: Alfa Aesar™ B2514703

Quantity 1g

 View more versions of this product

Catalog No. AAB2514703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1569-08-0
    Molecular Formula C12H15NO
    Formula Weight 189.26
    MDL Number MFCD00102140
    Synonym 2-Benzyl-4,4-dimethyl-4,5-dihydrooxazole
    Chemical Name or Material 2-Benzyl-4,4-dimethyl-2-oxazoline

    Specifications

    Formula Weight 189.26
    Molecular Formula C12H15NO
    Health Hazard 1 Warning
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00102140
    CAS 1569-08-0
    Synonym 2-Benzyl-4,4-dimethyl-4,5-dihydrooxazole
    RTECS Number RP6798960
    Chemical Name or Material 2-Benzyl-4,4-dimethyl-2-oxazoline
    Assay 95%

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit