Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Benzyl-4,4-dimethyl-2-oxazoline, 95%

Click to view available options
Quantity:
1g
5g
1569-08-0
C12H15NO
189.258
MFCD00102140
GPBCTHQIXXYBHQ-UHFFFAOYSA-N
2-benzyl-4,4-dimethyl-4,5-dihydrooxazole,2-benzyl-4,4-dimethyl-2-oxazoline,2-benzyl-4,4-dimethyl-4,5-dihydro-1,3-oxazole,oxazole, 4,5-dihydro-4,4-dimethyl-2-phenylmethyl,4,4-dimethyl-2-phenylmethyl-4,5-dihydrooxazole,acmc-20a6ng,2-benzyl-4,4-dimethyl-5h-oxazole,4,4-dimethyl-2-benzyl-1,3-oxazoline,oxazole,4,5-dihydro-4,4-dimethyl-2-phenylmethyl
98710
2-benzyl-4,4-dimethyl-5H-1,3-oxazole
CC1(COC(=N1)CC2=CC=CC=C2)C
1569-08-0
C12H15NO
189.258
MFCD00102140
GPBCTHQIXXYBHQ-UHFFFAOYSA-N
2-benzyl-4,4-dimethyl-4,5-dihydrooxazole,2-benzyl-4,4-dimethyl-2-oxazoline,2-benzyl-4,4-dimethyl-4,5-dihydro-1,3-oxazole,oxazole, 4,5-dihydro-4,4-dimethyl-2-phenylmethyl,4,4-dimethyl-2-phenylmethyl-4,5-dihydrooxazole,acmc-20a6ng,2-benzyl-4,4-dimethyl-5h-oxazole,4,4-dimethyl-2-benzyl-1,3-oxazoline,oxazole,4,5-dihydro-4,4-dimethyl-2-phenylmethyl
98710
2-benzyl-4,4-dimethyl-5H-1,3-oxazole
CC1(COC(=N1)CC2=CC=CC=C2)C
Quantity 5g
Formula Weight 189.26
Percent Purity 95%
Chemical Name or Material 2-Benzyl-4,4-dimethyl-2-oxazoline

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : RP6798960