Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Benzylaminopyridine, 98%

Click to view available options
Quantity:
50g
250g
1000g
6935-27-9
C12H12N2
184.242
MFCD00006251
WYHXNQXDQQMTQI-UHFFFAOYSA-N
2-benzylaminopyridine,2-pyridinamine, n-phenylmethyl,2-benzylamino pyridine,pyridine, 2-benzylamino,n-benzyl-2-pyridinamine,benzyl-2-pyridylamine,enamine_005854,cbmicro_010916,n-2-pyridyl benzylamine,acmc-209o7g
23362
N-benzylpyridin-2-amine
C1=CC=C(C=C1)CNC2=CC=CC=N2
6935-27-9
C12H12N2
184.242
MFCD00006251
WYHXNQXDQQMTQI-UHFFFAOYSA-N
2-benzylaminopyridine,2-pyridinamine, n-phenylmethyl,2-benzylamino pyridine,pyridine, 2-benzylamino,n-benzyl-2-pyridinamine,benzyl-2-pyridylamine,enamine_005854,cbmicro_010916,n-2-pyridyl benzylamine,acmc-209o7g
23362
N-benzylpyridin-2-amine
C1=CC=C(C=C1)CNC2=CC=CC=N2
Boiling Point 182°C (12mmHg)
Flash Point 153°C (307°F)
Quantity 250g
Melting Point 93°C to 97°C
Beilstein 134309
Formula Weight 184.24
Percent Purity 98%
Chemical Name or Material 2-Benzylaminopyridine

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : US2700000

TSCA : TSCA