Alfa Aesar™ 2-Benzylidene-1-tetralone, 97%

Manufacturer: Alfa Aesar™ B2469906

Quantity 5g

 View more versions of this product

Catalog No. AAB2469906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6261-32-1
    Molecular Formula C17H14O
    Formula Weight 234.3
    MDL Number MFCD00019675
    Beilstein 476209
    Synonym 2-Benzylidene-1,2,3,4-tetrahydronaphthalen-1-one
    Chemical Name or Material 2-Benzylidene-1-tetralone

    Specifications

    Formula Weight 234.3
    Melting Point 103° to 105°C
    Quantity 5g
    Molecular Formula C17H14O
    MDL Number MFCD00019675
    CAS 6261-32-1
    Beilstein 476209
    Synonym 2-Benzylidene-1,2,3,4-tetrahydronaphthalen-1-one
    Chemical Name or Material 2-Benzylidene-1-tetralone
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit