Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Benzylphenol, 98+%

Click to view available options
Quantity:
10g
50g
250g
28994-41-4
C13H12O
184.238
MFCD00002242
CDMGNVWZXRKJNS-UHFFFAOYSA-N
2-hydroxydiphenylmethane,o-benzylphenol,phenol, 2-phenylmethyl,benzylphenol,2-benzyl-phenol,2-phenylmethyl phenol,alpha-phenyl-o-cresol,o-cresol, .alpha.-phenyl,unii-sf555bl24o,2-benzyl phenol
24216
2-benzylphenol
C1=CC=C(C=C1)CC2=CC=CC=C2O
28994-41-4
C13H12O
184.238
MFCD00002242
CDMGNVWZXRKJNS-UHFFFAOYSA-N
2-hydroxydiphenylmethane,o-benzylphenol,phenol, 2-phenylmethyl,benzylphenol,2-benzyl-phenol,2-phenylmethyl phenol,alpha-phenyl-o-cresol,o-cresol, .alpha.-phenyl,unii-sf555bl24o,2-benzyl phenol
24216
2-benzylphenol
C1=CC=C(C=C1)CC2=CC=CC=C2O
Melting Point 50°C to 54°C
Quantity 10g
Flash Point >110°C (230°F)
Boiling Point 310°C to 312°C
Beilstein 2045725
Merck Index 14,1142
Formula Weight 184.24
Percent Purity ≥98%
Chemical Name or Material 2-Benzylphenol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA