Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Benzylthiazolidine-4-carboxylic acid, 97%

Click to view available options
Quantity:
250mg
1g
5g
50739-30-5
C11H13NO2S
223.29
MFCD09936775
SNFSDSLKMQQXDU-UHFFFAOYSA-N
2-benzylthiazolidine-4-carboxylic acid,2-benzyl-thiazolidine-4-carboxylic acid,2-benzyl-4-thiazolidinecarboxylic acid
4285155
2-benzyl-1,3-thiazolidine-4-carboxylic acid
C1C(NC(S1)CC2=CC=CC=C2)C(=O)O
50739-30-5
C11H13NO2S
223.29
MFCD09936775
SNFSDSLKMQQXDU-UHFFFAOYSA-N
2-benzylthiazolidine-4-carboxylic acid,2-benzyl-thiazolidine-4-carboxylic acid,2-benzyl-4-thiazolidinecarboxylic acid
4285155
2-benzyl-1,3-thiazolidine-4-carboxylic acid
C1C(NC(S1)CC2=CC=CC=C2)C(=O)O
Quantity 1g
Formula Weight 223.29
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Benzylthiazolidine-4-carboxylic acid

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures