Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Benzylthio-5-nitrobenzonitrile, 97%

Click to view available options
Quantity:
1g
5g
175135-67-8
C14H10N2O2S
270.306
MFCD00067808
UVENNUQTKADSQA-UHFFFAOYSA-N
2-benzylthio-5-nitrobenzonitrile,2-benzylsulfanyl-5-nitrobenzonitrile,acmc-20aoo6,maybridge1_001743,benzonitrile,5-nitro-2-phenylmethyl thio,5-nitro-2-phenylmethylthio benzenecarbonitrile
706765
2-benzylsulfanyl-5-nitrobenzonitrile
C1=CC=C(C=C1)CSC2=C(C=C(C=C2)[N+](=O)[O-])C#N
175135-67-8
C14H10N2O2S
270.306
MFCD00067808
UVENNUQTKADSQA-UHFFFAOYSA-N
2-benzylthio-5-nitrobenzonitrile,2-benzylsulfanyl-5-nitrobenzonitrile,acmc-20aoo6,maybridge1_001743,benzonitrile,5-nitro-2-phenylmethyl thio,5-nitro-2-phenylmethylthio benzenecarbonitrile
706765
2-benzylsulfanyl-5-nitrobenzonitrile
C1=CC=C(C=C1)CSC2=C(C=C(C=C2)[N+](=O)[O-])C#N
Quantity 5g
UN Number UN3439
Formula Weight 270.31
Percent Purity 97%
Chemical Name or Material 2-Benzylthio-5-nitrobenzonitrile

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.