Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-BFI hydrochloride, Tocris Bioscience™

Catalog Number p-4783161
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
89196-95-2
C11H11ClN2O
222.672
RFNFFVDVGWOSNZ-UHFFFAOYSA-N
2-2-benzofuranyl-2-imidazoline hydrochloride, 2-bfi hydrochloride, 2-benzofuran-2-yl-2-imidazoline hydrochloride, 2-1-benzofuran-2-yl-4,5-dihydro-1h-imidazole hydrochloride, 2-bfi hydrochloride hplc, 2-2-benzo b furanyl-2-imidazoline hydrochloride, 2-1-benzofuran-2-yl-4,5-dihydro-1h-imidazole,hydrochloride, 2-benzofuran-2-yl-4,5-dihydro-1h-imidazole hydrochloride, 2-1-benzofuran-2-yl-4,5-dihydro-1h-imidazole-hydrogen chloride 1/1
45073465
2-(1-benzofuran-2-yl)-4,5-dihydro-1H-imidazole;hydrochloride
C1CN=C(N1)C2=CC3=CC=CC=C3O2.Cl
This item is not returnable. View return policy
89196-95-2
C11H11ClN2O
222.672
RFNFFVDVGWOSNZ-UHFFFAOYSA-N
2-2-benzofuranyl-2-imidazoline hydrochloride, 2-bfi hydrochloride, 2-benzofuran-2-yl-2-imidazoline hydrochloride, 2-1-benzofuran-2-yl-4,5-dihydro-1h-imidazole hydrochloride, 2-bfi hydrochloride hplc, 2-2-benzo b furanyl-2-imidazoline hydrochloride, 2-1-benzofuran-2-yl-4,5-dihydro-1h-imidazole,hydrochloride, 2-benzofuran-2-yl-4,5-dihydro-1h-imidazole hydrochloride, 2-1-benzofuran-2-yl-4,5-dihydro-1h-imidazole-hydrogen chloride 1/1
45073465
2-(1-benzofuran-2-yl)-4,5-dihydro-1H-imidazole;hydrochloride
C1CN=C(N1)C2=CC3=CC=CC=C3O2.Cl

Potent, selective I2 ligand. Putative agonist

High affinity I2 ligand (Ki = 9.8 nM). Putative I2 agonist; potentiates morphine antinociception. Produces ipsiversive rotational behavior in rats with a full 6-OHDA lesion of the nigrostriatal tract.
Quantity 10 mg
Formula Weight 222.67
Percent Purity >98%
Chemical Name or Material 2-BFI hydrochloride
Recommended Storage Desiccate at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit