Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Biphenylyl Glycidyl Ether 95%, ACROS Organics™

Click to view available options
Quantity:
5g
7144-65-2
C15H14O2
226.28
MFCD00046989
DNVXWIINBUTFEP-UHFFFAOYNA-N
2-biphenylyl glycidyl ether, opp-g, 3-2-xenyloxy propylene oxide, 2-1,1'-biphenyl-2-yloxy methyl oxirane, ccris 2065, 3-2-biphenyloxy propylene oxide, 3-2-xenolyl-1,2-epoxypropane, biphenyl-2-yl 2,3-epoxypropyl ether, ether, 2-biphenylyl 2,3-epoxypropyl, oxirane, 2-1,1'-biphenyl-2-yloxy methyl
97875
2-({[1,1'-biphenyl]-2-yloxy}methyl)oxirane
C(OC1=CC=CC=C1C1=CC=CC=C1)C1CO1
7144-65-2
C15H14O2
226.28
MFCD00046989
DNVXWIINBUTFEP-UHFFFAOYNA-N
2-biphenylyl glycidyl ether, opp-g, 3-2-xenyloxy propylene oxide, 2-1,1'-biphenyl-2-yloxy methyl oxirane, ccris 2065, 3-2-biphenyloxy propylene oxide, 3-2-xenolyl-1,2-epoxypropane, biphenyl-2-yl 2,3-epoxypropyl ether, ether, 2-biphenylyl 2,3-epoxypropyl, oxirane, 2-1,1'-biphenyl-2-yloxy methyl
97875
2-({[1,1'-biphenyl]-2-yloxy}methyl)oxirane
C(OC1=CC=CC=C1C1=CC=CC=C1)C1CO1
Quantity 5g
CAS Min % 94.0
CAS Max % 100.0
Beilstein 17, IV, 995
Formula Weight 226.27
Chemical Name or Material 2-Biphenylyl glycidyl ether, 95%

EINECSNumber : 230-451-0

RTECSNumber : RR0356000

TSCA : TSCA