Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino)benzaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
250mg
1202865-21-1
C37H51O3P
574.78
MFCD15143663
PAWZHFURVJBYGV-UHFFFAOYSA-N
2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphino benzaldehyde,2-bis 3,5-di-t-butyl-4-methoxyphenyl phosphino benzaldehyde,2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphanyl benzaldehyde,acmc-20ao6l,2-bis 3,5-di-t-butyl-4-methoxyphenyl phosphino benzaldehyde, min
71310909
2-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphanylbenzaldehyde
CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=CC=CC=C2C=O)C3=CC(=C(C(=C3)C(C)(C)C)OC)C(C)(C)C
1202865-21-1
C37H51O3P
574.78
MFCD15143663
PAWZHFURVJBYGV-UHFFFAOYSA-N
2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphino benzaldehyde,2-bis 3,5-di-t-butyl-4-methoxyphenyl phosphino benzaldehyde,2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphanyl benzaldehyde,acmc-20ao6l,2-bis 3,5-di-t-butyl-4-methoxyphenyl phosphino benzaldehyde, min
71310909
2-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphanylbenzaldehyde
CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=CC=CC=C2C=O)C3=CC(=C(C(=C3)C(C)(C)C)OC)C(C)(C)C
Quantity 250mg
Color Yellow
Melting Point 123.0°C to 124.5°C
Infrared Spectrum Authentic
NMR (1H-NMR) Consistent with structure, (31P-NMR) Consistent with structure, NMR (19F-NMR), Consistent with structure
Elemental Analysis 74.22 to 80.41% C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 574.78
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 2-(Bis(3, 5-di-tert-Butyl-4-methoxyphenyl)phosphino)benzaldehyde
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning