Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Bis(3,5-dimethylphenyl)phosphino)benzaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
250mg
669091-00-3
C23H23OP
346.41
MFCD15143662
PQIAXRGPKDEYLW-UHFFFAOYSA-N
2-bis 3,5-dimethylphenyl phosphino benzaldehyde,2-bis 3,5-dimethylphenyl phosphanyl benzaldehyde,acmc-20ao6m,2-bis 3,5-dimethylphenyl phosphino benzaldehyde,min
71310677
2-bis(3,5-dimethylphenyl)phosphanylbenzaldehyde
CC1=CC(=CC(=C1)P(C2=CC=CC=C2C=O)C3=CC(=CC(=C3)C)C)C
669091-00-3
C23H23OP
346.41
MFCD15143662
PQIAXRGPKDEYLW-UHFFFAOYSA-N
2-bis 3,5-dimethylphenyl phosphino benzaldehyde,2-bis 3,5-dimethylphenyl phosphanyl benzaldehyde,acmc-20ao6m,2-bis 3,5-dimethylphenyl phosphino benzaldehyde,min
71310677
2-bis(3,5-dimethylphenyl)phosphanylbenzaldehyde
CC1=CC(=CC(=C1)P(C2=CC=CC=C2C=O)C3=CC(=CC(=C3)C)C)C
NMR (1H-NMR) Consistent with structure, (31P-NMR) Consistent with structure, NMR (19F-NMR), Consistent with structure
Color Yellow
Elemental Analysis 76.56 to 82.94% C
Quantity 250mg
Melting Point 109.5°C to 111.0°C
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 346.41
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 2-(Bis(3, 5-Dimethylphenyl)phosphino)benzaldehyde
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning