Learn More
Accela Chembio IncSupplier Diversity Partner 2-(BOC-AMINO)-4-METHYLPEN 1G
Manufacturer:  Accela Chembio IncSupplier Diversity Partner SY0063611G
Encompass_Preferred
2-(boc-amino)-4-methylpentylamine, 1g, 1186663-67-1, MFCD12828717, 95+%, Shelf Life: 900 Days, Moisture Sensitive/nitrogen Or Argon/+4
Catalog No. 50-251-071