Alfa Aesar™ 2-(Boc-amino)-5-cyanopyridine, 97%

Manufacturer: Alfa Aesar™ H5007903

Quantity 1g

 View more versions of this product

Catalog No. AAH5007903

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 902837-44-9
    Molecular Formula C11H13N3O2
    Formula Weight 219.24
    MDL Number MFCD07781163
    Synonym 6-(Boc-amino)nicotinonitrile; 6-(Boc-amino)pyridine-3-carbonitrile
    Chemical Name or Material 2-(Boc-amino)-5-cyanopyridine

    Specifications

    Formula Weight 219.24
    Molecular Formula C11H13N3O2
    Melting Point 170° to 175°C
    MDL Number MFCD07781163
    Quantity 1g
    CAS 902837-44-9
    Synonym 6-(Boc-amino)nicotinonitrile; 6-(Boc-amino)pyridine-3-carbonitrile
    Health Hazard 2 GHS H Statement
    H302-H319
    Harmful if swallowed.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280f-P305+P351+P338
    Wear protective clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Health Hazard 1 Warning
    Chemical Name or Material 2-(Boc-amino)-5-cyanopyridine
    Assay 97%

    GHS H Statement
    H302-H319
    Harmful if swallowed.
    Causes serious eye irritation.

    GHS P Statement
    P280f-P305+P351+P338
    Wear protective clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit