Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(BOC-Amino)ethanethiol, 97%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
67385-09-5
C7H15NO2S
177.27
MFCD00274335
GSJJCZSHYJNRPN-UHFFFAOYSA-N
2-boc-amino ethanethiol,boc-cysteamine,tert-butyl n-2-mercaptoethyl carbamate,tert-butyl n-2-sulfanylethyl carbamate,2-bocamino ethanethiol,tert-butyl 2-mercaptoethyl carbamate,carbamic acid, 2-mercaptoethyl-, 1,1-dimethylethyl ester,n-boc-aet,tert-butoxy-n-2-sulfanylethyl carboxamide,n-boc-cysteamine
3017761
tert-butyl N-(2-sulfanylethyl)carbamate
CC(C)(C)OC(=O)NCCS
67385-09-5
C7H15NO2S
177.27
MFCD00274335
GSJJCZSHYJNRPN-UHFFFAOYSA-N
2-boc-amino ethanethiol,boc-cysteamine,tert-butyl n-2-mercaptoethyl carbamate,tert-butyl n-2-sulfanylethyl carbamate,2-bocamino ethanethiol,tert-butyl 2-mercaptoethyl carbamate,carbamic acid, 2-mercaptoethyl-, 1,1-dimethylethyl ester,n-boc-aet,tert-butoxy-n-2-sulfanylethyl carboxamide,n-boc-cysteamine
3017761
tert-butyl N-(2-sulfanylethyl)carbamate
CC(C)(C)OC(=O)NCCS
Specific Gravity 1.049
Boiling Point 68.0°C (0.3 mmHg)
Color Colorless to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5mL
Refractive Index 1.4730 to 1.4770
Density 1.0490g/mL
Flash Point 109°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula (CH3)3COC(O)NH(CH2)2SH
Formula Weight 177.27
Physical Form Viscous Liquid or Low Melting Solid
Percent Purity 97%
Chemical Name or Material 2-(BOC-Amino)ethanethiol
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning