Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Boc-amino)ethyl bromide, 96%

Manufacturer:  Alfa Aesar™ H55211MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH55211MD


  Check Availability
Add to cart

Specifications

Description

Applications
N-t-Boc-2-bromoethylamine is a building block for preparing fluorinated spacers having nucleophilic and electrophilic termini.

Solubility
Soluble in Chloroform, Dichloromethane and Ethyl Acetate.

Notes
Store at -20°C. Reactive with oxidizing agents.
Description & Specifications

Chemical Identifiers

39684-80-5
BrCH2CH2NHCO2C(CH3)3
MFCD02683428
tert-Butyl N-(2-bromoethyl)carbamate
C7H14BrNO2
224.1
2325117
2-(Boc-amino)ethyl bromide

Specifications

39684-80-5
30° to 32°C
224.1
250mg
tert-Butyl N-(2-bromoethyl)carbamate
96%
C7H14BrNO2
BrCH2CH2NHCO2C(CH3)3
MFCD02683428
2325117
2-(Boc-amino)ethyl bromide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Recommended Storage : −30° to −10°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit