Alfa Aesar™ 2-(Boc-amino)ethyl bromide, 96%

Manufacturer: Alfa Aesar™ H5521106

Quantity 5g

 View more versions of this product

Catalog No. AAH5521106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 39684-80-5
    Molecular Formula C7H14BrNO2
    Linear Formula BrCH2CH2NHCO2C(CH3)3
    Formula Weight 224.1
    MDL Number MFCD02683428
    Beilstein 2325117
    Synonym tert-Butyl N-(2-bromoethyl)carbamate
    Chemical Name or Material 2-(Boc-amino)ethyl bromide

    Specifications

    Formula Weight 224.1
    Molecular Formula C7H14BrNO2
    Linear Formula BrCH2CH2NHCO2C(CH3)3
    Quantity 5g
    Beilstein 2325117
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 30° to 32°C
    MDL Number MFCD02683428
    CAS 39684-80-5
    Synonym tert-Butyl N-(2-bromoethyl)carbamate
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage −30° to −10°C
    Chemical Name or Material 2-(Boc-amino)ethyl bromide
    Assay 96%

    Description

    Applications
    N-t-Boc-2-bromoethylamine is a building block for preparing fluorinated spacers having nucleophilic and electrophilic termini.

    Solubility
    Soluble in Chloroform, Dichloromethane and Ethyl Acetate.

    Notes
    Store at -20°C. Reactive with oxidizing agents.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    −30° to −10°C

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit