Alfa Aesar™ 2-(Boc-amino)pyridine, 95%

Manufacturer: Alfa Aesar™ H30741MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH30741MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 38427-94-0
    Molecular Formula C10H14N2O2
    Formula Weight 194.23
    MDL Number MFCD03411622
    Synonym 2-(tert-Butoxycarbonylamino)pyridine
    Chemical Name or Material 2-(Boc-amino)pyridine

    Specifications

    Formula Weight 194.23
    Molecular Formula C10H14N2O2
    Melting Point 92° to 94°C
    MDL Number MFCD03411622
    Quantity 250mg
    CAS 38427-94-0
    Synonym 2-(tert-Butoxycarbonylamino)pyridine
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 2-(Boc-amino)pyridine
    Assay 95%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit