Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-(3-fluorophenyl)ethan-1-one, 97%, Thermo Scientific™

Catalog Number SPB06322EA
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
53631-18-8
C8H6BrFO
217.04
MFCD00109756
ITAQNNGDCNFGID-UHFFFAOYSA-N
2-bromo-1-3-fluorophenyl ethan-1-one, 2-bromo-1-3-fluorophenyl ethanone, 3-fluorophenacyl bromide, 2-bromo-3'-fluoroacetophenone, ethanone, 2-bromo-1-3-fluorophenyl, 3'-fluorophenacyl bromide, pubchem13417, acmc-20ao30, alpha-bromo-3'-fluoroacetophenone
2737451
2-bromo-1-(3-fluorophenyl)ethanone
FC1=CC=CC(=C1)C(=O)CBr
53631-18-8
C8H6BrFO
217.04
MFCD00109756
ITAQNNGDCNFGID-UHFFFAOYSA-N
2-bromo-1-3-fluorophenyl ethan-1-one, 2-bromo-1-3-fluorophenyl ethanone, 3-fluorophenacyl bromide, 2-bromo-3'-fluoroacetophenone, ethanone, 2-bromo-1-3-fluorophenyl, 3'-fluorophenacyl bromide, pubchem13417, acmc-20ao30, alpha-bromo-3'-fluoroacetophenone
2737451
2-bromo-1-(3-fluorophenyl)ethanone
FC1=CC=CC(=C1)C(=O)CBr
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 217.04
Percent Purity 97%
Chemical Name or Material 2-bromo-1-(3-fluorophenyl)ethan-1-one
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit