Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-(3-phenylisoxazol-5-yl)ethan-1-one 95%, Maybridge

Click to view available options
Quantity:
1g
Packaging:
Amber Glass Bottle
14731-14-7
C11H8BrNO2
266.094
XTBXGZOVSCTNEC-UHFFFAOYSA-N
5-bromoacetyl-3-phenylisoxazole,2-bromo-1-3-phenylisoxazol-5-yl ethan-1-one,2-bromo-1-3-phenylisoxazol-5-yl ethanone,2-bromo-1-3-phenyl-1,2-oxazol-5-yl ethanone,2-bromo-1-3-phenyl-1,2-oxazol-5-yl ethan-1-one,2-bromo-1-3-phenyl-isoxazol-5-yl-ethanone,ethanone, 2-bromo-1-3-phenyl-5-isoxazolyl,pubchem8717,buttpark 43\57-72
2735532
2-bromo-1-(3-phenyl-1,2-oxazol-5-yl)ethanone
C1=CC=C(C=C1)C2=NOC(=C2)C(=O)CBr
14731-14-7
C11H8BrNO2
266.094
XTBXGZOVSCTNEC-UHFFFAOYSA-N
5-bromoacetyl-3-phenylisoxazole,2-bromo-1-3-phenylisoxazol-5-yl ethan-1-one,2-bromo-1-3-phenylisoxazol-5-yl ethanone,2-bromo-1-3-phenyl-1,2-oxazol-5-yl ethanone,2-bromo-1-3-phenyl-1,2-oxazol-5-yl ethan-1-one,2-bromo-1-3-phenyl-isoxazol-5-yl-ethanone,ethanone, 2-bromo-1-3-phenyl-5-isoxazolyl,pubchem8717,buttpark 43\57-72
2735532
2-bromo-1-(3-phenyl-1,2-oxazol-5-yl)ethanone
C1=CC=C(C=C1)C2=NOC(=C2)C(=O)CBr
Packaging Amber Glass Bottle
Quantity 1g
Assay Percent Range 95%
Formula Weight 266.09
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(3-phenylisoxazol-5-yl)ethan-1-one