Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1,3-thiazole-4-carboxylic acid, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
5198-88-9
C4H2BrNO2S
208.03
BEGREHRAUWCAHV-UHFFFAOYSA-N
2-bromo-4-thiazolecarboxylic acid,2-bromothiazole-4-carboxylic acid,2-bromo-thiazole-4-carboxylic acid,4-thiazolecarboxylic acid, 2-bromo,2-bromo-4-thiazolecarboxylicacid,2-bromothiazole-4-carboxylicacid,2-bromo-4-carboxy-1,3-thiazole,pubchem22900,acmc-1avkr,2-bromo-4-carboxythiazole
2763209
2-bromo-1,3-thiazole-4-carboxylic acid
C1=C(N=C(S1)Br)C(=O)O
5198-88-9
C4H2BrNO2S
208.03
BEGREHRAUWCAHV-UHFFFAOYSA-N
2-bromo-4-thiazolecarboxylic acid,2-bromothiazole-4-carboxylic acid,2-bromo-thiazole-4-carboxylic acid,4-thiazolecarboxylic acid, 2-bromo,2-bromo-4-thiazolecarboxylicacid,2-bromothiazole-4-carboxylicacid,2-bromo-4-carboxy-1,3-thiazole,pubchem22900,acmc-1avkr,2-bromo-4-carboxythiazole
2763209
2-bromo-1,3-thiazole-4-carboxylic acid
C1=C(N=C(S1)Br)C(=O)O
Melting Point 204.0°C to 205.5°C
Quantity 5g
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 208.03
Percent Purity 98%
Chemical Name or Material 2-Bromo-1,3-thiazole-4-carboxylic acid

GHS H Statement Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. Harmful if inhaled. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning