Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-1,4-dimethoxybenzene, 98%

Click to view available options
Quantity:
10g
50g
250g
25245-34-5
C8H9BrO2
217.062
MFCD00008355
DWCGNRKFLRLWCJ-UHFFFAOYSA-N
1-bromo-2,5-dimethoxybenzene,benzene, 2-bromo-1,4-dimethoxy,2,5-dimethoxybromobenzene,pubchem19785,2-bromo-4-methoxyanisole,acmc-1cby3,ksc490m9p,1,4-dimethoxy-2-bromobenzene,1-bromo-2, 5-dimethoxybenzene,dwcgnrkflrlwcj-uhfffaoysa
91353
2-bromo-1,4-dimethoxybenzene
COC1=CC(=C(C=C1)OC)Br
25245-34-5
C8H9BrO2
217.062
MFCD00008355
DWCGNRKFLRLWCJ-UHFFFAOYSA-N
1-bromo-2,5-dimethoxybenzene,benzene, 2-bromo-1,4-dimethoxy,2,5-dimethoxybromobenzene,pubchem19785,2-bromo-4-methoxyanisole,acmc-1cby3,ksc490m9p,1,4-dimethoxy-2-bromobenzene,1-bromo-2, 5-dimethoxybenzene,dwcgnrkflrlwcj-uhfffaoysa
91353
2-bromo-1,4-dimethoxybenzene
COC1=CC(=C(C=C1)OC)Br
Applications
2-Bromo-1,4-dimethoxybenzene has been used in preparation of S-(-)-2-[N-(trifluoroacetyl)amino]-1-(2,5-dimethoxy-4- bromophenyl)-1-propanone, 1,4-bis{4-[2-(4-pyridyl)ethenyl]phenyl}butadiyne and 1,4-bis{2,5-dimethoxy-4-[2-(4-pyridyl)ethenyl]phenyl}butadiyne and 1-bromo-4-iodo-2,5-dimethoxybenzene. It can be used as a chemical redox shuttle additive in Lithium ion batteries for repeated overcharge and overdischarge protection.

Solubility
Insoluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Light Sensitive.
Refractive Index 1.57
Density 1.441
Flash Point >110°C (230°F)
Boiling Point 260°C to 262°C
Quantity 250g
Beilstein 2441884
Solubility Information Insoluble in water.
Formula Weight 217.07
Percent Purity 98%
Chemical Name or Material 2-Bromo-1,4-dimethoxybenzene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 246-756-7

TSCA : Yes

Recommended Storage : Ambient temperatures