Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-[4-(trifluoromethyl)phenyl]ethan-1-one, Tech., Maybridge

Click to view available options
Quantity:
1g
10g
Packaging:
Amber Glass Bottle
383-53-9
C9H6BrF3O
267.045
MFCD00126489
HEMROKPXTCOASZ-UHFFFAOYSA-N
2-bromo-1-4-trifluoromethyl phenyl ethan-1-one,4-trifluoromethyl phenacyl bromide,2-bromo-4'-trifluoromethyl acetophenone,2-bromo-1-4-trifluoromethyl phenyl ethanone,2-bromo-4'-trifluoromethyl acetophenon,4-trifluoromethylphenacyl bromide,ethanone, 2-bromo-1-4-trifluoromethyl phenyl,4'-trifluoromethyl phenacyl bromide,2-bromo-4'-trifluoromethylacetophenone
321979
2-bromo-1-[4-(trifluoromethyl)phenyl]ethanone
C1=CC(=CC=C1C(=O)CBr)C(F)(F)F
383-53-9
C9H6BrF3O
267.045
MFCD00126489
HEMROKPXTCOASZ-UHFFFAOYSA-N
2-bromo-1-4-trifluoromethyl phenyl ethan-1-one,4-trifluoromethyl phenacyl bromide,2-bromo-4'-trifluoromethyl acetophenone,2-bromo-1-4-trifluoromethyl phenyl ethanone,2-bromo-4'-trifluoromethyl acetophenon,4-trifluoromethylphenacyl bromide,ethanone, 2-bromo-1-4-trifluoromethyl phenyl,4'-trifluoromethyl phenacyl bromide,2-bromo-4'-trifluoromethylacetophenone
321979
2-bromo-1-[4-(trifluoromethyl)phenyl]ethanone
C1=CC(=CC=C1C(=O)CBr)C(F)(F)F
Packaging Amber Glass Bottle
Quantity 1g
Assay Percent Range Tech.
Linear Formula CF3C6H4C(O)CH2Br
Formula Weight 267.04
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Bromo-1-[4-(trifluoromethyl)phenyl]ethan-1-one

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger