Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Bromo-1-butene, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
2-Bromo-1-butene is used to produce 2-Ethyl-acrylic acid. It acts as a photolytic precursor for 1-buten-2-yl.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-1-butene is used to produce 2-Ethyl-acrylic acid. It acts as a photolytic precursor for 1-buten-2-yl.
Solubility
Soluble in ethanol, acetone and benzene. Not miscible in water.
Notes
Store away from air. Store under dry inert gas. Incompatible with oxidizing agents and air.
2-Bromo-1-butene is used to produce 2-Ethyl-acrylic acid. It acts as a photolytic precursor for 1-buten-2-yl.
Solubility
Soluble in ethanol, acetone and benzene. Not miscible in water.
Notes
Store away from air. Store under dry inert gas. Incompatible with oxidizing agents and air.
Specifications
Refractive Index | 1.4525 |
Quantity | 5 g |
UN Number | UN1993 |
Synonym | 2-bromo-1-butene, 1-butene, 2-bromo, sjphabimsp@, 2-bromobutene, 2-bromo-l-butene, 1-butene,2-bromo |
Solubility Information | Soluble in ethanol,acetone and benzene. Not miscible in water. |
Formula Weight | 135 |
Percent Purity | 97% |
Chemical Name or Material | 2-Bromo-1-butene |
Safety and Handling
Hazard Category | H225 |
Hazard Statement | GHS H Statement H224 Extremely flammable liquid and vapour. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
EINECSNumber | 245-412-3 |
TSCA | Yes |
Recommended Storage | Store at -20°C |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction