Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-1-butene, 97%

Click to view available options
Quantity:
5g
25g
23074-36-4
C4H7Br
135.004
MFCD00039178
HQMXRIGBXOFKIU-UHFFFAOYSA-N
2-bromo-1-butene,1-butene, 2-bromo,sjphabimsp@,2-bromobutene,2-bromo-l-butene,1-butene,2-bromo
89990
2-bromobut-1-ene
CCC(=C)Br
23074-36-4
C4H7Br
135.004
MFCD00039178
HQMXRIGBXOFKIU-UHFFFAOYSA-N
2-bromo-1-butene,1-butene, 2-bromo,sjphabimsp@,2-bromobutene,2-bromo-l-butene,1-butene,2-bromo
89990
2-bromobut-1-ene
CCC(=C)Br
Applications
2-Bromo-1-butene is used to produce 2-Ethyl-acrylic acid. It acts as a photolytic precursor for 1-buten-2-yl.

Solubility
Soluble in ethanol, acetone and benzene. Not miscible in water.

Notes
Store away from air. Store under dry inert gas. Incompatible with oxidizing agents and air.
Refractive Index 1.4525
Quantity 5g
UN Number UN1993
Solubility Information Soluble in ethanol,acetone and benzene. Not miscible in water.
Formula Weight 135
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-butene

GHS H Statement
H224
Extremely flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 245-412-3

TSCA : Yes

Recommended Storage : Store at -20°C